accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 5122-82-7

IUPAC Name: 1-(adamantan-1-yl)-2-bromoethan-1-one
Synonyms: 1-(adamantan-1-yl)-2-bromoethanone
Molecular Weight (g/mol): 257.17
Molecular Formula: C12H17BrO
InChi Key: KWCDIRFSULAMOC-UHFFFAOYSA-N
SMILES: BrCC(=O)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results