accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COBO

CAS RN 5123-13-7

IUPAC Name: 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane
Synonyms: 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 190.05
Molecular Formula: C11H15BO2
InChi Key: FQMSNYZDFMWLGC-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)