Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Trimethylsilyl)Methanesulfonate

Catalog No. 6429610MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-296-10MLF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 64-296-10MLF Supplier Sigma Aldrich Supplier No. 6429610MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant or reagent involved in:Reduction of esters to ethers using boron difluoride triflate etherateElimination of C(8)-functional groups via regioselective dehydrationDealkylsilylation of alumazene derivativesSilylating agent

Specifications

CAS 5123-13-7
Melting Point 62°C to 66°C
Molecular Formula C11H15BO2
Refractive Index n20/D 1.422; n20/D 1.422
Linear Formula C11H15BO2
MDL Number MFCD05663851
Quantity 5 g
Synonym Methanesulfonic acid trimethylsilyl ester; Trimethylsilyl mesylate
Molecular Weight (g/mol) 190.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.