accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NONONH3C

CAS RN 51235-04-2

IUPAC Name: 3-cyclohexyl-6-(dimethylamino)-1-methyl-1,2,3,4-tetrahydro-1,3,5-triazine-2,4-dione
Synonyms: hexazinone velpar weed killer epa pesticide chemical code 107201 velpar hexazinone, (14)C-labeled caswell no. 271AA (14)c-labeled hexazinone gridball hexazinone, (14)c-labeled (14)C-labeled hexazinone
Molecular Weight (g/mol): 252.32
Molecular Formula: C12H20N4O2
InChi Key: CAWXEEYDBZRFPE-UHFFFAOYSA-N
SMILES: CN(C)C1=NC(=O)N(C2CCCCC2)C(=O)N1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results