accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51260-39-0

IUPAC Name: 4-methyl-1,3-dioxolan-2-one
Synonyms: propylene carbonate arconate 5000 propylene carbonate preparation 4-methyldioxalone-2 propylene glycol cyclic carbonate propylenester kyseliny uhlicite carbonic acid, cyclic propylene ester carbonic acid, cyclic propylene ether 1,2-propylene carbonate dipropylene carbonate
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILES: CC1COC(=O)O1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results