accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNONH(E/Z)

CAS RN 51285-05-3

IUPAC Name: 1-(1,2-dihydropyrazin-2-ylidene)-1-nitrosomethanamine
Synonyms: 1-nitroso-1-(1H-pyrazin-2-ylidene)methanamine
Molecular Weight (g/mol): 138.13
Molecular Formula: C5H6N4O
InChi Key: IXCAPURAZXLMLQ-UHFFFAOYSA-N
SMILES: NC(N=O)=C1NC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results