accessibility menu, dialog, popup

UserName

Viewing profile as user:
CoOOO

CAS RN 513-79-1

IUPAC Name: λ²-cobalt(2+) carbonate
Synonyms: cobalt(II) carbonate cobaltous carbonate
Molecular Weight (g/mol): 118.94
Molecular Formula: CCoO3
InChi Key: ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
SMILES: [Co++].[O-]C([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)