Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Carbonate, MP Biomedicals

Catalog No. ICN223075 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN223075 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN223075 Supplier MP Biomedicals, Inc Supplier No. 0522307550

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 513-79-1
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00010940
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv
PubChem CID 10565
IUPAC Name λ²-cobalt(2+) carbonate
SMILES [Co++].[O-]C([O-])=O

Specifications

Quantity 100 g
Formula Weight 118.92
Chemical Name or Material Cobaltous Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.