accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5131-66-8

IUPAC Name: 1-butoxypropan-2-ol
Synonyms: 2-propanol, 1-butoxy- propylene glycol butyl ether
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: RWNUSVWFHDHRCJ-UHFFFAOYNA-N
SMILES: CCCCOCC(C)O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results