CAS RN 51317-66-9
IUPAC Name:
4-(4-isothiocyanatophenyl)morpholine
Synonyms:
4-(4-isothiocyanatophenyl)morpholine
Molecular Weight (g/mol):
220.29
Molecular Formula:
C11H12N2OS
InChi Key:
AXUXRZZYZBZQAR-UHFFFAOYSA-N
SMILES:
S=C=NC1=CC=C(C=C1)N1CCOCC1
1
–
2
of
2
results
1
4-Morpholinophenyl isothiocyanate, Thermo Scientific™
CAS: 51317-66-9 Molecular Formula: C11H12N2OS Molecular Weight (g/mol): 220.29 InChI Key: AXUXRZZYZBZQAR-UHFFFAOYSA-N PubChem CID: 224862 IUPAC Name: 4-(4-isothiocyanatophenyl)morpholine SMILES: C1COCCN1C2=CC=C(C=C2)N=C=S
2
Sigma Aldrich 4-morpholinophenyl isothiocyanate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.