Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-morpholinophenyl isothiocyanate

Catalog No. CDS014671 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014671 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014671 Supplier Sigma Aldrich Supplier No. CDS014671100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethyl chloromalonate (dimethyl 2-chloromalonate) is a dialkyl 2-substituted malonate
  • Carbanions of dimethyl chloromalonate are reported to react with naphthoquinone derivatives, via vicarious nucleophilic substitution or oxidative nucleophilic substitution of hydrogen processes
  • Dimethyl chloromalonate is reported to react with hydroquinone and monosubstituted hydroquinones to afford 2-oxido-benzo[b]furan derivatives
  • Synthesis of dimethyl 2-chloromalonate, via chlorination of dimethyl malonate has been reported.dimethyl chloromalonate is suitable for use in the synthesis of 4,5,6-trichloropyrimidine
  • It may be used in the stereoselective synthesis of highly functionalized nitrocyclopropanes
  • It may be used in one-pot synthesis of tetra-alkoxycarbonylallylidenetriphenylphosphoranes

Specifications

CAS 51317-66-9
Molecular Formula C11H12N2OS
Linear Formula C11H12N2OS
MDL Number MFCD00047409
Quantity 100 g
Molecular Weight (g/mol) 220.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.