accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51323-43-4

IUPAC Name: [3-(bromomethyl)phenyl]boronic acid
Synonyms: 3-(bromomethyl)phenylboronic acid
Molecular Weight (g/mol): 214.85
Molecular Formula: C7H8BBrO2
InChi Key: ATRFDLFMCLYROQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(CBr)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)