Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Bromomethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B38145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3814-5G 5 g
B38141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3814-5G Supplier TCI America Supplier No. B38145G
Add to Cart
Add to Cart

Specifications

CAS 51323-43-4
Melting Point 216°C
Color White-Yellow
Molecular Formula C7H8BBrO2
MDL Number MFCD01632207
Quantity 5 g
UN Number 3261
Synonym 3-bromomethylphenylboronic acid, 3-bromomethyl phenylboronic acid, 3-bromomethyl benzeneboronic acid, 3-bromomethyl phenyl boronic acid, 3-bromomethylphenyl boronic acid
InChI Key ATRFDLFMCLYROQ-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)CBr)(O)O
IUPAC Name [3-(bromomethyl)phenyl]boronic acid
Molecular Weight (g/mol) 214.853
PubChem CID 2773281
Formula Weight 214.85
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.