accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51338-27-3

IUPAC Name: methyl 2-[4-(2,4-dichlorophenoxy)phenoxy]propanoate
Synonyms: diclofop methyl dichlorfop-methyl hoegrass dichlofopmethyl hoelon 3EC dichlordiphenprop diclofop-methyl (+-)-diclofop-methyl methyl diclofop methyldiclofop
Molecular Weight (g/mol): 341.18
Molecular Formula: C16H14Cl2O4
InChi Key: BACHBFVBHLGWSL-UHFFFAOYNA-N
SMILES: COC(=O)C(C)OC1=CC=C(OC2=CC=C(Cl)C=C2Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results