accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51359-78-5

IUPAC Name: 4-(bromomethyl)benzaldehyde
Synonyms: 4-(bromomethyl)benzaldehyde
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: XYPVBKDHERGKJG-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results