Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Bromomethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog No. MO07312DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07312DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07312DE Supplier Thermo Scientific Chemicals Supplier No. MO07312DE
Add to Cart

Chemical Identifiers

CAS 51359-78-5
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
InChI Key XYPVBKDHERGKJG-UHFFFAOYSA-N
PubChem CID 11206421
IUPAC Name 4-(bromomethyl)benzaldehyde
SMILES C1=CC(=CC=C1CBr)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde
Formula Weight 199.05
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.