accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5137-55-3

IUPAC Name: methyltrioctylazanium chloride
Synonyms: aliquat 336
Molecular Weight (g/mol): 404.16
Molecular Formula: C25H54ClN
InChi Key: XKBGEWXEAPTVCK-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)