Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyltrioctylammonium Chloride

Catalog No. 6948550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-485-50G 50 g
69-485-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 69-485-50G Supplier Sigma Aldrich Supplier No. 6948550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyltrioctylammonium chloride can be used:As a catalyst in the synthesis of acridine dione derivatives from aromatic aldehyde, dimedone and amines under ultrasonic irradiations.As a catalyst in the synthesis of extended π-systems using aromatic aldehydes and methyldiazines.As a component of a catalytic system used in the Suzuki-Miyaura cross-coupling reaction of a variety of aryl and heteroaryl chlorides in H2O.

Specifications

CAS 5137-55-3
Molecular Formula C25H54ClN
Linear Formula [CH3(CH2)6CH2]3N(Cl)CH3
MDL Number MFCD00011862
Quantity 50 g
Synonym Trioctylmethylammonium chloride
Molecular Weight (g/mol) 404.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.