accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 514-10-3

IUPAC Name: (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Synonyms: abietic acid abietic acid, barium salt abietic acid, ammonium salt cobalt salt abietic acid zinc salt abietic acid kyselina abietova sodium salt abietic acid abietic acid, potassium salt palladium (+2) salt abietic acid abietic acid, calcium salt
Molecular Weight (g/mol): 302.46
Molecular Formula: C20H30O2
InChi Key: RSWGJHLUYNHPMX-ONCXSQPRSA-N
SMILES: CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)