Learn More
Abietic acid, tech. 75%
Description
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.
Solubility
Soluble in alcohols, acetone and ethers
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. Sensitive to air.
Specifications
Specifications
| Quantity | 100 g |
| Odor | Characteristic |
| Sensitivity | Air sensitive |
| Beilstein | 2221451 |
| Merck Index | 14,7 |
| Solubility Information | Soluble in alcohols,acetone and ethers |
| Formula Weight | 302.45 |
| Percent Purity | 75% |
| Grade | Technical |
| Chemical Name or Material | Abietic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 208-178-3 |
| RTECSNumber | TP8580000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.