accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH

CAS RN 5140-03-4

IUPAC Name: 2-[4-(2-hydroxyethyl)phenyl]ethan-1-ol
Synonyms: 2-[4-(2-hydroxyethyl)phenyl]ethanol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: LBZZJNPUANNABV-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(CCO)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results