Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(2-hydroxyethyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B22955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22955G Supplier TCI America Supplier No. B22955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 5140-03-4
Melting Point 88°C
Color White
Boiling Point 193°C
Molecular Formula C10H14O2
Quantity 5 g
MDL Number MFCD00666548
Synonym 1,4-Benzenediethanol, 1,4-Phenylenediethanol
InChI Key LBZZJNPUANNABV-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCO)CCO
IUPAC Name 2-[4-(2-hydroxyethyl)phenyl]ethanol
Molecular Weight (g/mol) 166.22
PubChem CID 5013205
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-hydroxyethyl)benzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.