accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBrN

CAS RN 5140-72-7

IUPAC Name: 2-bromo-6-chloropyridine
Synonyms: 2-bromo-6-chloropyridine
Molecular Weight (g/mol): 192.44
Molecular Formula: C5H3BrClN
InChi Key: JWTZSVLLPKTZJP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Br)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)