accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSSSSCH3

CAS RN 514188-77-3

IUPAC Name: 5-decyl-5'-{5'-decyl-[2,2'-bithiophen]-5-yl}-2,2'-bithiophene
Synonyms: 5-decyl-5'-{5'-decyl-[2,2'-bithiophen]-5-yl}-2,2'-bithiophene
Molecular Weight (g/mol): 611.04
Molecular Formula: C36H50S4
InChi Key: IYCHXHNXTBDCNT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(CCCCCCCCCC)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results