accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BiH3CCH3

CAS RN 5142-75-6

IUPAC Name: tris(4-methylphenyl)bismuthane
Synonyms: tris(4-methylphenyl)bismuthane
Molecular Weight (g/mol): 482.38
Molecular Formula: C21H21Bi
InChi Key: XIECNHSGPKDTNW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)[Bi](C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results