Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-p-tolylbismuthine 98.0+%, TCI America™
SDP

Catalog No. T22611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22611G Supplier TCI America Supplier No. T22611G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5142-75-6
Molecular Formula C21H21Bi
Molecular Weight (g/mol) 482.38
MDL Number MFCD00123272
InChI Key XIECNHSGPKDTNW-UHFFFAOYSA-N
Synonym Tris(4-methylphenyl)bismuthine, Tris(p-tolyl)bismuthine
PubChem CID 340259
IUPAC Name tris(4-methylphenyl)bismuthane
SMILES CC1=CC=C(C=C1)[Bi](C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 482.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tri-p-tolylbismuthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.