accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51446-62-9

IUPAC Name: 2-amino-3-({[2,3-bis(octadecanoyloxy)propoxy](hydroxy)phosphoryl}oxy)propanoic acid
Synonyms: 2-amino-3-{[2,3-bis(octadecanoyloxy)propoxy(hydroxy)phosphoryl]oxy}propanoic acid
Molecular Weight (g/mol): 792.09
Molecular Formula: C42H82NO10P
InChi Key: TZCPCKNHXULUIY-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCC(N)C(O)=O)OC(=O)CCCCCCCCCCCCCCCCC
Grade 
  • (2)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)