Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphatidyl-L-serine, ∽98%, MP Biomedicals™

Catalog No. ICN10096950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10096950 50 mg
ICN10096910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10096950 Supplier MP Biomedicals, Inc Supplier No. 0210096950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51446-62-9 | C42H82NO10P | 792.089 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51446-62-9
Molecular Formula C42H82NO10P
Molecular Weight (g/mol) 792.089
InChI Key TZCPCKNHXULUIY-RGULYWFUSA-N
Synonym distearoyl phosphatidylserine, 2s-2-amino-3-r-2,3-bis stearoyloxy propoxy hydroxy phosphoryl oxy propanoic acid, phosphatidylserine 36:0, dioctadecanoylphosphatidylserine, pser 36:0, 1,2-dioctadecanoyl-sn-glycero-3-phosphoserine, phosphatidylserine 18:0/18:0, l-serine, 2,3-bis 1-oxooctadecyl oxy propyl ester, dihydrogen phosphate ester , r, 1,2-distearoyl-sn-glycero-3-phospho-l-serine, l-alpha-phosphatidyl-l-serine
PubChem CID 9547096
ChEBI CHEBI:84519
IUPAC Name (2S)-2-amino-3-[[(2R)-2,3-di(octadecanoyloxy)propoxy]-hydroxyphosphoryl]oxypropanoic acid
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCC(C(=O)O)N)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Color Yellow
Assay Percent Range ∽ 98%
Quantity 50 mg
Formula Weight 788
Percent Purity ∼98%
Grade Biochemical
Physical Form Liquid
Chemical Name or Material Phosphatidyl-L-serine
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.