accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5149-85-9

IUPAC Name: bis(4-(diethylamino)benzene-1-diazonium); tetrachlorozincdiuide
Synonyms: bis(4-(diethylamino)benzenediazonium); tetrachlorozincdiuide
Molecular Weight (g/mol): 563.7
Molecular Formula: C20H32Cl4N6Zn
InChi Key: IGMATLBHQBHWSV-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.[Zn--].CCN(CC)C1=CC=C(C=C1)[N+]#N.CCN(CC)C1=CC=C(C=C1)[N+]#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results