Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Diazo-N,N-diethylaniline, ZnCl2, MP Biomedicals

Catalog No. ICN210679 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210679 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ICN210679 Supplier MP Biomedicals, Inc Supplier No. 0521067950

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5149-85-9
Molecular Formula C20H28Cl4N6Zn
Molecular Weight (g/mol) 559.666
InChI Key KSUPIOGMHIUCNV-UHFFFAOYSA-J
Synonym benzenediazonium, 4-diethylamino-, t-4-tetrachlorozincate 2-2:1, diazo 517p, 4-diazo-n,n-diethylaniline chloride zinc chloride, p-diazo-n,n'-diethylaniline chloride zinc chloride, p-diazonium-n,n-diethylaniline, zinc chloride 2:1, bis 4-diethylamino benzenediazonium tetrachlorozincate, 4-diethylamino benzenediazonium tetrachlorozincate 2:1, 4-diethylaminobenzenediazonium chloride zinc chloride 2:1, bis 4-diethylamino benzenediazonium tetrachlorozincate 2-, benzenediazonium, p-diethylamino-, tetrachlorozincate 2-2:1
PubChem CID 78827
IUPAC Name 4-(diethylamino)benzenediazonium;tetrachlorozinc(2-)
SMILES CCN(CC)C1=CC=C(C=C1)[N+]#N.CCN(CC)C1=CC=C(C=C1)[N+]#N.Cl[Zn-2](Cl)(Cl)Cl

Specifications

Quantity 100 g
Chemical Name or Material p-Diazo-N, N-diethylaniline ZnCl2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.