accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)CH3(R)CH3OH(R)(R)H3COHCH2

CAS RN 515-03-7

IUPAC Name: (1R,2R,8aS)-1-[(3R)-3-hydroxy-3-methylpent-4-en-1-yl]-2,5,5,8a-tetramethyl-decahydronaphthalen-2-ol
Synonyms: sclareol
Molecular Weight (g/mol): 308.51
Molecular Formula: C20H36O2
InChi Key: XVULBTBTFGYVRC-FFADBYAMSA-N
SMILES: C[C@@](O)(CC[C@H]1[C@](C)(O)CCC2C(C)(C)CCC[C@]12C)C=C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)