accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFFFFFFFFFFFFFFF

CAS RN 51502-45-5

IUPAC Name: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
Molecular Weight (g/mol): 478.11
Molecular Formula: C10H3F17O2
InChi Key: CFNCFMKWDPDIPT-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results