Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Heptadecafluorononanoate 95.0+%, TCI America™
SDP

Catalog No. M19155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19155G 5 g
M191525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19155G Supplier TCI America Supplier No. M19155G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51502-45-5
Molecular Formula C10H3F17O2
Molecular Weight (g/mol) 478.105
MDL Number MFCD00153213
InChI Key CFNCFMKWDPDIPT-UHFFFAOYSA-N
Synonym methyl perfluorononanoate, methyl heptadecafluorononanoate, heptadecafluorononanoic acid methyl ester, acmc-209ku2, perfluorononanoic acid methyl ester, heptadecafluorononanoic acid, methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecakis fluoranyl nonanoate, nonanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, methyl ester, heptadecafluorononanoic acid methyl ester; methyl perfluorononanoate; perfluorononanoic acid methyl ester
PubChem CID 2775476
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 65°C
Quantity 5 g
Formula Weight 478.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Heptadecafluorononanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.