CAS RN 5153-24-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
227.33
Molecular Formula:
C4H8O5Zr
InChi Key:
ZMLDZZAIXBVKJU-UHFFFAOYSA-N
SMILES:
O=[Zr].CC(O)=O.CC(O)=O
1
–
1
of
1
results
1
Diacetoxyzirconium(IV) Oxide (ca. 20% in Water), TCI America™
CAS: 5153-24-2 Molecular Formula: C4H6O5Zr Synonym: Bis(acetato-O)oxozirconium(IV)