Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetoxyzirconium(IV) Oxide (ca. 20% in Water), TCI America™
SDP

Catalog No. Z0030500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0030500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z0030500G Supplier TCI America Supplier No. Z0030500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5153-24-2
Molecular Formula C4H6O5Zr
Synonym Bis(acetato-O)oxozirconium(IV)

Specifications

Color Yellow
Quantity 500 g
Formula Weight 225.31
Physical Form Liquid
Chemical Name or Material Diacetoxyzirconium(IV) Oxide (ca. 20% in Water)
EINECSNumber (2)-2199&(9)-0480
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.