CAS RN 5153-73-1
IUPAC Name:
4-(2-nitroethenyl)benzonitrile
Synonyms:
4-(2-nitroethenyl)benzonitrile
Molecular Weight (g/mol):
174.16
Molecular Formula:
C9H6N2O2
InChi Key:
CVWPMONBGNBYCJ-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C=CC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
trans-4-(2-Nitroethenyl)benzonitrile 98.0+%, TCI America™
CAS: 5153-73-1 Molecular Formula: C9H6N2O2 Molecular Weight (g/mol): 174.159 MDL Number: MFCD00176174 InChI Key: CVWPMONBGNBYCJ-AATRIKPKSA-N Synonym: trans-4-(2-Nitrovinyl)benzonitrile PubChem CID: 819894 IUPAC Name: 4-[(E)-2-nitroethenyl]benzonitrile SMILES: C1=CC(=CC=C1C=C[N+](=O)[O-])C#N