Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(2-Nitroethenyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. N07081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N07081G Supplier TCI America Supplier No. N07081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5153-73-1
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00176174
InChI Key CVWPMONBGNBYCJ-AATRIKPKSA-N
Synonym trans-4-(2-Nitrovinyl)benzonitrile
PubChem CID 819894
IUPAC Name 4-[(E)-2-nitroethenyl]benzonitrile
SMILES C1=CC(=CC=C1C=C[N+](=O)[O-])C#N

Specifications

Melting Point 190°C
Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 174.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(2-Nitroethenyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.