accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)OHOF

CAS RN 51543-40-9

IUPAC Name: (2R)-2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Synonyms: flurbiprofen sodium furbiprofen flurbiprofeno [inn-spanish] flurbiprofenum (R)-flurbiprofen flurbiprofene [inn-french] FLP tarenflurbil R-flurbiprofen flurizan
Molecular Weight (g/mol): 244.27
Molecular Formula: C15H13FO2
InChi Key: SYTBZMRGLBWNTM-SNVBAGLBSA-N
SMILES: C[C@@H](C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results