Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-Flurbiprofen, 97%

Catalog No. AC434490010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC434490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434490010 Supplier Thermo Scientific Chemicals Supplier No. 434490010
Add to Cart

CAS: 51543-40-9 | C15H13FO2 | 244.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51543-40-9
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00869714
InChI Key SYTBZMRGLBWNTM-SNVBAGLBSA-N
Synonym tarenflurbil, r-flurbiprofen, flurizan, r-2-flurbiprofen, unii-501w00oowa, r-2-flubiprofen, r---flurbiprofen, --2r-2-2-fluorobiphenyl-4-yl propanoic acid, r---2-fluoro-alpha-methyl-4-biphenylacetic acid
PubChem CID 92337
ChEBI CHEBI:38666
IUPAC Name (2R)-2-(3-fluoro-4-phenylphenyl)propanoic acid
SMILES CC(C1=CC(=C(C=C1)C2=CC=CC=C2)F)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 106.0°C to 110.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 1 g
Merck Index 15,9196
Specific Rotation Condition − 40.00 (19.10°C c=1 in CHCl3)
Specific Rotation -40
Formula Weight 244.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-Flurbiprofen
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with pl
EINECSNumber 257-264-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.