accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)NO(S)(R)ONHHOOOSiH3CCH3CH3CH3CH3

CAS RN 51549-36-1

IUPAC Name: N-{1-[(2R,3R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Synonyms: N-{1-[(2R,3R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-2-oxopyrimidin-4-yl}benzamide
Molecular Weight (g/mol): 461.59
Molecular Formula: C22H31N3O6Si
InChi Key: CXSIUDLYCLLBGH-DLVXIWMQSA-N
SMILES: CC(C)(C)[Si](C)(C)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results