Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxycytidine, 98+%, Thermo Scientific™

Catalog No. AAH52296MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52296MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52296MD Supplier Thermo Scientific Chemicals Supplier No. H52296MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51549-36-1
Packaging Glass bottle
Molecular Formula C22H31N3O5Si
Quantity 250mg
MDL Number MFCD04972284
Formula Weight 445.59
Percent Purity ≥98%
Chemical Name or Material N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxycytidine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.