accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5157-08-4

IUPAC Name: 6-methyl-2,3,4,5-tetrahydropyridazin-3-one
Synonyms: 6-methyl-4,5-dihydro-2H-pyridazin-3-one
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: VOTFXESXPPEARL-UHFFFAOYSA-N
SMILES: CC1=NNC(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results