Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dihydro-6-methyl-3(2H)-pyridazinone 98.0+%, TCI America™
SDP

Catalog No. D421625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D421625G 25 g
D42165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D421625G Supplier TCI America Supplier No. D421625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5157-08-4
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.13
MDL Number MFCD00006455
InChI Key VOTFXESXPPEARL-UHFFFAOYSA-N
Synonym 6-methyl-4,5-dihydropyridazin-3 2h-one, 4,5-dihydro-6-methylpyridazin-3 2h-one, 6-methyl-2,3,4,5-tetrahydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dihydro-6-methyl, 4,5-dihydro-6-methyl-3 2h-pyridazinone, 6-methyl-4,5-dihydro-2h-pyridazin-3-one, 6-methyl-4,5-dihydro-3 2h-pyridazinone, wln: t6nmv futj f1, 6-methyl-2,4,5-trihydropyridazin-3-one, 3 2h-pyridazinone,5-dihydro-6-methyl
PubChem CID 78834
IUPAC Name 3-methyl-4,5-dihydro-1H-pyridazin-6-one
SMILES CC1=NNC(=O)CC1

Specifications

Melting Point 81°C
Color White
Quantity 25 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dihydro-6-methyl-3(2H)-pyridazinone
RTECSNumber UR6375000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.