accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2ONaClNONONClO

CAS RN 51580-86-0

IUPAC Name: sodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide dihydrate
Synonyms: dichloroisocyanuric acid
Molecular Weight (g/mol): 255.97
Molecular Formula: C3H4Cl2N3NaO5
InChi Key: PYILKOIEIHHYGD-UHFFFAOYSA-M
SMILES: O.O.[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)