accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHClCl

CAS RN 51619-56-8

IUPAC Name: 1-[(2,4-dichlorophenyl)methyl]piperazine
Synonyms: 1-[(2,4-dichlorophenyl)methyl]piperazine
Molecular Weight (g/mol): 245.15
Molecular Formula: C11H14Cl2N2
InChi Key: VYKXBWXIOSLDQR-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(CN2CCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results