accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51628-12-7

IUPAC Name: 2-(4-iodophenyl)acetonitrile
Synonyms: 2-(4-iodophenyl)acetonitrile
Molecular Weight (g/mol): 243.05
Molecular Formula: C8H6IN
InChi Key: PNXWQTYSBFGIFD-UHFFFAOYSA-N
SMILES: IC1=CC=C(CC#N)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results