Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenylacetonitrile, 97%

Catalog No. AAL1511906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1511906 5 g
AAL1511903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1511906 Supplier Thermo Scientific Chemicals Supplier No. L1511906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51628-12-7 | C8H6IN | 243.047 g/mol

4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51628-12-7
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.047
MDL Number MFCD00060306
InChI Key PNXWQTYSBFGIFD-UHFFFAOYSA-N
Synonym 4-iodophenylacetonitrile, 2-4-iodophenyl acetonitrile, 4-iodobenzyl cyanide, 4-iodophenyl acetonitrile, benzeneacetonitrile, 4-iodo, p-iodobenzylcyanide, pubchem19992, acmc-1asuk, 4-cyanomethyl phenyl iodide, 4-iodo-phenyl-acetonitrile
PubChem CID 142866
IUPAC Name 2-(4-iodophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)I

Specifications

Melting Point 52°C to 54°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3439
Beilstein 1934479
Sensitivity Light sensitive
Solubility Information Insoluble in water
Formula Weight 243.05
Percent Purity 97%
Chemical Name or Material 4-Iodophenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.