accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNNH2

CAS RN 51644-96-3

IUPAC Name: tert-butyl N-(5-aminopentyl)carbamate
Synonyms: tert-butyl N-(5-aminopentyl)carbamate
Molecular Weight (g/mol): 202.3
Molecular Formula: C10H22N2O2
InChi Key: DPLOGSUBQDREOU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCCN
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)