accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CO

CAS RN 51673-84-8

IUPAC Name: 2,2-dimethoxyacetaldehyde
Synonyms: 2,2-dimethoxyacetaldehyde
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: OGFKTAMJLKHRAZ-UHFFFAOYSA-N
SMILES: COC(OC)C=O
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)