accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 517-23-7

IUPAC Name: 3-acetyloxolan-2-one
Synonyms: 3-acetyloxolan-2-one
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: OMQHDIHZSDEIFH-UHFFFAOYNA-N
SMILES: CC(=O)C1CCOC1=O
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)